2,3-Dichloro-5-(prop-2-yn-1-yloxy)pyridine ≥95%

Supplier: AOBChem USA

363975-5G 363975-25G
77493-570EA 1484.23 USD
77493-570 77503-334
2,3-Dichloro-5-(prop-2-yn-1-yloxy)pyridine ≥95%
2,3-Dichloro-5-(prop-2-yn-1-yloxy)pyridine

Precaución: For research use only

Formula: C₈H₅Cl₂NO
CAS Number: 2322698-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR