Methyl 5-hydroxy-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 3-hydroxy-5-Methylbenzoate

36360-5G 36360-25G
77493-566EA 430.54 USD
77493-566 77503-330
Methyl 5-hydroxy-3-methylbenzoate ≥97%
Methyl 5-hydroxy-3-methylbenzoate

Precaución: For research use only

Formula: C₉H₁₀O₃
MW: 166.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11007825
CAS Number: 2615-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR