1-(3-Bromophenyl)-2,2,2-trifluoroethanol ≥95%

Supplier: AOBChem USA

Warning

36305-5G 36305-25G
77493-552EA 719.97 USD
77493-552 77503-318
1-(3-Bromophenyl)-2,2,2-trifluoroethanol ≥95%
1-(3-Bromophenyl)-2,2,2-trifluoroethanol

Precaución: For research use only

Formula: C₈H₆BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD16067907
CAS Number: 446-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR