2-Fluoro-4-(methoxymethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

36210-5G 36210-25G
77493-548EA 891.98 USD
77493-548 77503-314
2-Fluoro-4-(methoxymethyl)benzaldehyde ≥97%
2-Fluoro-4-(methoxymethyl)benzaldehyde

Precaución: For research use only

Formula: C₉H₉FO₂
MDL Number: MFCD34756006
CAS Number: 1782055-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR