Cyclopropyl(2,3-dichloro-6-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

361088-1G 361088-500MG 361088-5G 361088-25G
77470-416EA 415.09 USD
77470-416 77483-084 77493-520 77503-296
Cyclopropyl(2,3-dichloro-6-fluorophenyl)methanol ≥95%
Cyclopropyl(2,3-dichloro-6-fluorophenyl)methanol

Precaución: For research use only

Formula: C₁₀H₉Cl₂FO
Storage Temperature: Ambient
CAS Number: 3002509-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR