1-(2,3-Dichloro-6-fluorophenyl)-2-ethylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

361084-1G 361084-500MG 361084-5G 361084-25G
77470-412EA 415.09 USD
77470-412 77483-080 77493-516 77503-292
1-(2,3-Dichloro-6-fluorophenyl)-2-ethylbutan-1-ol ≥95%
1-(2,3-Dichloro-6-fluorophenyl)-2-ethylbutan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₅Cl₂FO
MW: 265.151 g/mol
Storage Temperature: Ambient
CAS Number: 3002424-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR