Butyl(2,3-dichloro-6-fluorophenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

361080-1G 361080-500MG 361080-5G 361080-25G
77470-408EA 415.09 USD
77470-408 77483-076 77493-512 77503-288
Butyl(2,3-dichloro-6-fluorophenyl)sulfane ≥95%
Butyl(2,3-dichloro-6-fluorophenyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₁Cl₂FS
Storage Temperature: Ambient
CAS Number: 3002446-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR