1-(2-Chloro-6-fluoro-3-methylphenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

361047-1G 361047-500MG 361047-5G 361047-25G
77470-400EA 415.09 USD
77470-400 77483-068 77493-504 77503-282
1-(2-Chloro-6-fluoro-3-methylphenyl)butan-1-ol ≥95%
1-(2-Chloro-6-fluoro-3-methylphenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄ClFO
MW: 216.68 g/mol
Storage Temperature: Ambient
CAS Number: 3002424-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR