(2-Chloro-6-fluoro-3-methylphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

361045-1G 361045-500MG 361045-5G 361045-25G
77470-396EA 415.09 USD
77470-396 77483-064 77493-500 77503-278
(2-Chloro-6-fluoro-3-methylphenyl)(cyclopropyl)methanol ≥95%
(2-Chloro-6-fluoro-3-methylphenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₁H₁₂ClFO
Storage Temperature: Ambient
CAS Number: 3002513-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR