1-(2-Chloro-6-fluoro-3-methylphenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

361042-1G 361042-500MG 361042-5G 361042-25G
77470-392EA 415.09 USD
77470-392 77483-060 77493-496 77503-274
1-(2-Chloro-6-fluoro-3-methylphenyl)propan-1-ol ≥95%
1-(2-Chloro-6-fluoro-3-methylphenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₂ClFO
MW: 202.653 g/mol
Storage Temperature: Ambient
CAS Number: 3002508-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR