1-(2-Chloro-6-fluoro-3-methylphenyl)-2-methylbutan-1-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
361039-1G
361039-500MG
361039-5G
361039-25G
77470-388EA
415.09
USD
77470-388
77483-056
77493-492
77503-270
1-(2-Chloro-6-fluoro-3-methylphenyl)-2-methylbutan-1-ol ≥95%
1-(2-Chloro-6-fluoro-3-methylphenyl)-2-methylbutan-1-ol
Precaución: For research use only
Formula:
C₁₂H₁₆ClFO MW: 230.706 g/mol Storage Temperature: Ambient |
CAS Number:
3002553-46-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...