(2-Chloro-6-fluoro-3-methylphenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

361034-1G 361034-500MG 361034-5G 361034-25G
77470-384EA 415.09 USD
77470-384 77483-052 77493-488 77503-266
(2-Chloro-6-fluoro-3-methylphenyl)(propyl)sulfane ≥95%
(2-Chloro-6-fluoro-3-methylphenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₁₀H₁₂ClFS
MW: 218.719 g/mol
Storage Temperature: Ambient
CAS Number: 3002526-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR