1-(2,6-Difluoro-3-iodophenyl)-3-methylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

361002-1G 361002-500MG 361002-5G 361002-25G
77470-380EA 415.09 USD
77470-380 77483-048 77493-484 77503-262
1-(2,6-Difluoro-3-iodophenyl)-3-methylbutan-1-ol ≥95%
1-(2,6-Difluoro-3-iodophenyl)-3-methylbutan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃F₂IO
MW: 326.122 g/mol
Storage Temperature: Ambient
CAS Number: 3002461-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR