Cyclopropyl(2,6-difluoro-3-iodophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

360987-1G 360987-500MG 360987-5G 360987-25G
77470-376EA 415.09 USD
77470-376 77483-044 77493-480 77503-258
Cyclopropyl(2,6-difluoro-3-iodophenyl)methanol ≥95%
Cyclopropyl(2,6-difluoro-3-iodophenyl)methanol

Precaución: For research use only

Formula: C₁₀H₉F₂IO
Storage Temperature: Ambient
CAS Number: 3002461-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR