3,5-Dichloro-2-isobutoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

360903-1G 360903-500MG 360903-5G 360903-25G
77470-372EA 415.09 USD
77470-372 77483-040 77493-476 77503-256
3,5-Dichloro-2-isobutoxybenzaldehyde ≥95%
3,5-Dichloro-2-isobutoxybenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₂Cl₂O₂
MW: 247.1177 g/mol
Storage Temperature: Ambient
CAS Number: 1153165-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR