1-(2-Chloro-6-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

360899-1G 360899-500MG 360899-5G 360899-25G
77470-368EA 397.58 USD
77470-368 77483-036 77493-472 77503-252
1-(2-Chloro-6-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2-Chloro-6-fluorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₄ClFO
MW: 216.6796 g/mol
Storage Temperature: Ambient
CAS Number: 1488765-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR