1-[4-(Cyclopentyloxy)phenyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

360513-1G 360513-500MG 360513-5G 360513-25G
77470-364EA 173.04 USD
77470-364 77483-032 77493-468 77503-248
1-[4-(Cyclopentyloxy)phenyl]ethanone ≥95%
1-[4-(Cyclopentyloxy)phenyl]ethanone

Precaución: For research use only

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Storage Temperature: Ambient
CAS Number: 857563-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR