(2-Chloro-6-fluorophenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

360348-1G 360348-500MG 360348-5G 360348-25G
77470-360EA 415.09 USD
77470-360 77483-028 77493-464 77503-244
(2-Chloro-6-fluorophenyl)(propyl)sulfane ≥95%
(2-Chloro-6-fluorophenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀ClFS
MW: 204.692 g/mol
Storage Temperature: Ambient
CAS Number: 3002480-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR