2,2-Dimethyl-1-(3-methyl-5-(trimethylsilyl)phenyl)propan-1-ol ≥95%

Supplier: AOBChem USA

Warning

360229-1G 360229-500MG 360229-5G 360229-25G
77470-356EA 415.09 USD
77470-356 77483-024 77493-460 77503-240
2,2-Dimethyl-1-(3-methyl-5-(trimethylsilyl)phenyl)propan-1-ol ≥95%
2,2-Dimethyl-1-(3-methyl-5-(trimethylsilyl)phenyl)propan-1-ol

Precaución: For research use only

Formula: C₁₅H₂₆OSi
MW: 250.452 g/mol
Storage Temperature: Ambient
CAS Number: 3002450-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR