2,2-Difluoro-1-(3-methyl-5-(trimethylsilyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

360219-1G 360219-500MG 360219-5G 360219-25G
77470-352EA 415.09 USD
77470-352 77483-020 77493-456 77503-236
2,2-Difluoro-1-(3-methyl-5-(trimethylsilyl)phenyl)ethanone ≥95%
2,2-Difluoro-1-(3-methyl-5-(trimethylsilyl)phenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₆F₂OSi
MW: 242.337 g/mol
Storage Temperature: Ambient
CAS Number: 3002472-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR