(3-(Ethylthio)-5-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

360212-1G 360212-500MG 360212-5G 360212-25G
77470-348EA 415.09 USD
77470-348 77483-016 77493-452 77503-232
(3-(Ethylthio)-5-methylphenyl)trimethylsilane ≥95%
(3-(Ethylthio)-5-methylphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₂H₂₀SSi
MW: 224.438 g/mol
Storage Temperature: Ambient
CAS Number: 3002507-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR