2-(2-(Benzyloxy)-3-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

360200-1G 360200-500MG 360200-5G 360200-25G
77470-344EA 415.09 USD
77470-344 77483-012 77493-448 77503-228
2-(2-(Benzyloxy)-3-methylphenyl)propan-2-ol ≥95%
2-(2-(Benzyloxy)-3-methylphenyl)propan-2-ol

Precaución: For research use only

Formula: C₁₇H₂₀O₂
MW: 256.339 g/mol
Storage Temperature: Ambient
CAS Number: 3002432-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR