1-(2-(Benzyloxy)-3-methylphenyl)-2-ethylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

360189-1G 360189-500MG 360189-5G 360189-25G
77470-340EA 415.09 USD
77470-340 77483-008 77493-444 77503-224
1-(2-(Benzyloxy)-3-methylphenyl)-2-ethylbutan-1-ol ≥95%
1-(2-(Benzyloxy)-3-methylphenyl)-2-ethylbutan-1-ol

Precaución: For research use only

Formula: C₂₀H₂₆O₂
MW: 298.419 g/mol
Storage Temperature: Ambient
CAS Number: 3002450-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR