1-(2-(Benzyloxy)-3-methylphenyl)-2-methylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

360186-1G 360186-500MG 360186-5G 360186-25G
77470-336EA 415.09 USD
77470-336 77483-004 77493-440 77503-220
1-(2-(Benzyloxy)-3-methylphenyl)-2-methylpropan-1-ol ≥95%
1-(2-(Benzyloxy)-3-methylphenyl)-2-methylpropan-1-ol

Precaución: For research use only

Formula: C₁₈H₂₂O₂
MW: 270.366 g/mol
Storage Temperature: Ambient
CAS Number: 3002432-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR