1-(2-(Benzyloxy)-3-methylphenyl)ethanol ≥95%

Supplier: AOBChem USA

Warning

360184-1G 360184-500MG 360184-5G
77470-332EA 497.49 USD
77470-332 77483-000 77493-436
1-(2-(Benzyloxy)-3-methylphenyl)ethanol ≥95%
1-(2-(Benzyloxy)-3-methylphenyl)ethanol

Precaución: For research use only

Formula: C₁₆H₁₈O₂
Storage Temperature: Ambient
CAS Number: 1226295-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR