1-(2-(Benzyloxy)-3-methylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

360180-1G 360180-500MG 360180-5G 360180-25G
77470-328EA 415.09 USD
77470-328 77482-996 77493-432 77503-214
1-(2-(Benzyloxy)-3-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(2-(Benzyloxy)-3-methylphenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₆H₁₃F₃O₂
MW: 294.268 g/mol
Storage Temperature: Ambient
CAS Number: 3002508-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR