1-(2-(Benzyloxy)-3-methylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

360173-1G 360173-500MG 360173-5G 360173-25G
77470-324EA 415.09 USD
77470-324 77482-992 77493-428 77503-210
1-(2-(Benzyloxy)-3-methylphenyl)-2,2-difluoroethanone ≥95%
1-(2-(Benzyloxy)-3-methylphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₆H₁₄F₂O₂
MW: 276.278 g/mol
Storage Temperature: Ambient
CAS Number: 3002424-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR