1-(3-Bromo-5-methylphenyl)-1-phenylethanol ≥95%

Supplier: AOBChem USA

Warning

360151-1G 360151-500MG 360151-5G 360151-25G
77470-320EA 415.09 USD
77470-320 77482-988 77493-424 77503-206
1-(3-Bromo-5-methylphenyl)-1-phenylethanol ≥95%
1-(3-Bromo-5-methylphenyl)-1-phenylethanol

Precaución: For research use only

Formula: C₁₅H₁₅BrO
Storage Temperature: Ambient
CAS Number: 3002507-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR