1-Bromo-4,5-dichloro-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

35904-1G 35904-500MG 35904-5G 35904-25G
77470-316EA 366.68 USD
77470-316 77482-984 77493-420 77503-202
1-Bromo-4,5-dichloro-2-iodobenzene ≥95%
1-Bromo-4,5-dichloro-2-iodobenzene

Precaución: For research use only

Formula: C₆H₂BrCl₂I
MW: 351.795 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672939
CAS Number: 289038-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR