α-Ethyl[1,1'-biphenyl]-2-methanol ≥95%

Supplier: AOBChem USA

Warning

358177-1G 358177-500MG 358177-5G
77470-312EA 487.19 USD
77470-312 77482-980 77493-416
α-Ethyl[1,1'-biphenyl]-2-methanol ≥95%
α-Ethyl[1,1'-biphenyl]-2-methanol

Precaución: For research use only

Formula: C₁₅H₁₆O
MW: 212.29 g/mol
Storage Temperature: Ambient
CAS Number: 875855-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR