(3-(cyclopropylmethoxy)phenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

357841-1G 357841-500MG 357841-5G 357841-25G
77470-304EA 415.09 USD
77470-304 77482-972 77493-408 77503-192
(3-(cyclopropylmethoxy)phenyl)(ethyl)sulfane ≥95%
(3-(cyclopropylmethoxy)phenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₆OS
Storage Temperature: Ambient
CAS Number: 3002544-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR