1-(6-phenylpyridin-3-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

357833-1G 357833-500MG 357833-5G 357833-25G
77470-300EA 415.09 USD
77470-300 77482-966 77493-402 77503-190
1-(6-phenylpyridin-3-yl)propan-1-one ≥95%
1-(6-phenylpyridin-3-yl)propan-1-one

Precaución: For research use only

Formula: C₁₄H₁₃NO
MW: 211.259 g/mol
Storage Temperature: Ambient
CAS Number: 2827065-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR