2-(6-phenylpyridin-3-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

357817-1G 357817-500MG 357817-5G 357817-25G
77470-296EA 415.09 USD
77470-296 77482-964 77493-400 77503-186
2-(6-phenylpyridin-3-yl)propan-2-ol ≥95%
2-(6-phenylpyridin-3-yl)propan-2-ol

Precaución: For research use only

Formula: C₁₄H₁₅NO
MW: 213.275 g/mol
Storage Temperature: Ambient
CAS Number: 1373310-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR