1-(4'-Butyl-[1,1'-biphenyl]-4-yl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

357777-1G 357777-500MG 357777-5G 357777-25G
77470-292EA 415.09 USD
77470-292 77482-960 77493-396 77503-182
1-(4'-Butyl-[1,1'-biphenyl]-4-yl)-2,2-dimethylpropan-1-ol ≥95%
1-(4'-Butyl-[1,1'-biphenyl]-4-yl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₂₁H₂₈O
MW: 296.446 g/mol
Storage Temperature: Ambient
CAS Number: 3002458-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR