1-(4'-Butyl-[1,1'-biphenyl]-4-yl)-2-methylbutan-1-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
357763-1G
357763-500MG
357763-5G
357763-25G
77470-288EA
415.09
USD
77470-288
77482-956
77493-392
77503-178
1-(4'-Butyl-[1,1'-biphenyl]-4-yl)-2-methylbutan-1-ol ≥95%
1-(4'-Butyl-[1,1'-biphenyl]-4-yl)-2-methylbutan-1-ol
Precaución: For research use only
Formula:
C₂₁H₂₈O MW: 296.446 g/mol Storage Temperature: Ambient |
CAS Number:
3002470-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...