(4'-butyl-[1,1'-biphenyl]-4-yl)(phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

357760-1G 357760-500MG 357760-5G 357760-25G
77470-284EA 415.09 USD
77470-284 77482-952 77493-388 77503-174
(4'-butyl-[1,1'-biphenyl]-4-yl)(phenyl)methanone ≥95%
(4'-butyl-[1,1'-biphenyl]-4-yl)(phenyl)methanone

Precaución: For research use only

Formula: C₂₃H₂₂O
Storage Temperature: Ambient
CAS Number: 3002449-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR