3-(2-Fluoro-5-iodophenyl)pentan-3-ol ≥95%

Supplier: AOBChem USA

Warning

357740-1G 357740-500MG 357740-5G 357740-25G
77470-280EA 415.09 USD
77470-280 77482-948 77493-384 77503-170
3-(2-Fluoro-5-iodophenyl)pentan-3-ol ≥95%
3-(2-Fluoro-5-iodophenyl)pentan-3-ol

Precaución: For research use only

Formula: C₁₁H₁₄FIO
MW: 308.131 g/mol
Storage Temperature: Ambient
CAS Number: 3002449-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR