(3-Chlorophenyl)(2-fluoro-5-iodophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

357738-1G 357738-500MG 357738-5G 357738-25G
77470-276EA 415.09 USD
77470-276 77482-944 77493-380 77503-166
(3-Chlorophenyl)(2-fluoro-5-iodophenyl)methanol ≥95%
(3-Chlorophenyl)(2-fluoro-5-iodophenyl)methanol

Precaución: For research use only

Formula: C₁₃H₉ClFIO
Storage Temperature: Ambient
CAS Number: 3002465-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR