2-Ethyl-1-(2-fluoro-5-iodophenyl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

357735-1G 357735-500MG 357735-5G 357735-25G
77470-272EA 415.09 USD
77470-272 77482-940 77493-376 77503-162
2-Ethyl-1-(2-fluoro-5-iodophenyl)butan-1-ol ≥95%
2-Ethyl-1-(2-fluoro-5-iodophenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆FIO
MW: 322.158 g/mol
Storage Temperature: Ambient
CAS Number: 3002449-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR