5-Fluoro-1-isopropoxy-3-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

35763-1G 35763-500MG 35763-5G 35763-25G
77470-268EA 81.37 USD
77470-268 77482-936 77493-372 77503-158
5-Fluoro-1-isopropoxy-3-(trifluoromethyl)benzene ≥95%
5-Fluoro-1-isopropoxy-3-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₁₀H₁₀F₄O
MW: 222.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23113806
CAS Number: 1395039-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR