Cyclohexyl(3-(methylthio)phenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

356925-1G 356925-500MG 356925-5G
77470-264EA 594.31 USD
77470-264 77482-932 77493-366
Cyclohexyl(3-(methylthio)phenyl)methanone ≥95%
Cyclohexyl(3-(methylthio)phenyl)methanone

Precaución: For research use only

Formula: C₁₄H₁₈OS
Storage Temperature: Ambient
CAS Number: 1926953-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR