2-Methyl-1-(3-(methylthio)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

356919-1G 356919-500MG 356919-5G 356919-25G
77470-260EA 324.45 USD
77470-260 77482-928 77493-362 77503-150
2-Methyl-1-(3-(methylthio)phenyl)propan-1-one ≥95%
2-Methyl-1-(3-(methylthio)phenyl)propan-1-one

Precaución: For research use only

Formula: C₁₁H₁₄OS
MW: 194.2932 g/mol
Storage Temperature: Ambient
CAS Number: 1352206-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR