(3-Isopropyl-4-methoxyphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

356624-1G 356624-500MG 356624-5G 356624-25G
77470-256EA 415.09 USD
77470-256 77482-924 77493-358 77503-146
(3-Isopropyl-4-methoxyphenyl)(phenyl)methanol ≥95%
(3-Isopropyl-4-methoxyphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₇H₂₀O₂
MW: 256.339 g/mol
Storage Temperature: Ambient
CAS Number: 3002447-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR