2-Chloro-2,2-difluoro-1-(3-isopropyl-4-methoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

356597-1G 356597-500MG 356597-5G 356597-25G
77470-252EA 415.09 USD
77470-252 77482-920 77493-354 77503-142
2-Chloro-2,2-difluoro-1-(3-isopropyl-4-methoxyphenyl)ethanone ≥95%
2-Chloro-2,2-difluoro-1-(3-isopropyl-4-methoxyphenyl)ethanone

Precaución: For research use only

Formula: C₁₂H₁₃ClF₂O₂
MW: 262.68 g/mol
Storage Temperature: Ambient
CAS Number: 3002513-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR