2,2,3,3,3-pentafluoro-1-(3-isopropyl-4-methoxyphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

356593-1G 356593-500MG 356593-5G 356593-25G
77470-248EA 415.09 USD
77470-248 77482-916 77493-350 77503-138
2,2,3,3,3-pentafluoro-1-(3-isopropyl-4-methoxyphenyl)propan-1-one ≥95%
2,2,3,3,3-pentafluoro-1-(3-isopropyl-4-methoxyphenyl)propan-1-one

Precaución: For research use only

Formula: C₁₃H₁₃F₅O₂
Storage Temperature: Ambient
CAS Number: 3002532-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR