2-Ethyl-1-(4'-isopropoxy-[1,1'-biphenyl]-4-yl)butan-1-ol ≥95%

Supplier: AOBChem USA

Warning

356543-1G 356543-500MG 356543-5G 356543-25G
77470-244EA 415.09 USD
77470-244 77482-912 77493-346 77503-134
2-Ethyl-1-(4'-isopropoxy-[1,1'-biphenyl]-4-yl)butan-1-ol ≥95%
2-Ethyl-1-(4'-isopropoxy-[1,1'-biphenyl]-4-yl)butan-1-ol

Precaución: For research use only

Formula: C₂₁H₂₈O₂
MW: 312.446 g/mol
Storage Temperature: Ambient
CAS Number: 3002423-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR