2,2,3,3,3-pentafluoro-1-(4'-isopropoxy-[1,1'-biphenyl]-4-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

356529-1G 356529-500MG 356529-5G 356529-25G
77470-240EA 415.09 USD
77470-240 77482-908 77493-342 77503-130
2,2,3,3,3-pentafluoro-1-(4'-isopropoxy-[1,1'-biphenyl]-4-yl)propan-1-one ≥95%
2,2,3,3,3-pentafluoro-1-(4'-isopropoxy-[1,1'-biphenyl]-4-yl)propan-1-one

Precaución: For research use only

Formula: C₁₈H₁₅F₅O₂
MW: 358.303 g/mol
Storage Temperature: Ambient
CAS Number: 3002461-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR