1-(4'-isopropoxy-[1,1'-biphenyl]-4-yl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

356526-1G 356526-500MG 356526-5G 356526-25G
77470-236EA 415.09 USD
77470-236 77482-904 77493-338 77503-126
1-(4'-isopropoxy-[1,1'-biphenyl]-4-yl)propan-1-one ≥95%
1-(4'-isopropoxy-[1,1'-biphenyl]-4-yl)propan-1-one

Precaución: For research use only

Formula: C₁₈H₂₀O₂
MW: 268.35 g/mol
Storage Temperature: Ambient
CAS Number: 1177230-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR