1-(3-Ethoxy-2-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

356470-1G 356470-500MG 356470-5G 356470-25G
77470-232EA 415.09 USD
77470-232 77482-900 77493-334 77503-122
1-(3-Ethoxy-2-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-Ethoxy-2-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₄H₂₂O₂
MW: 222.323 g/mol
Storage Temperature: Ambient
CAS Number: 3002484-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR