1-(3-Ethoxy-2-methylphenyl)-2-ethylbutan-1-ol ≥95%

Supplier: AOBChem USA

Warning

356451-1G 356451-500MG 356451-5G 356451-25G
77470-228EA 415.09 USD
77470-228 77482-896 77493-330 77503-118
1-(3-Ethoxy-2-methylphenyl)-2-ethylbutan-1-ol ≥95%
1-(3-Ethoxy-2-methylphenyl)-2-ethylbutan-1-ol

Precaución: For research use only

Formula: C₁₅H₂₄O₂
Storage Temperature: Ambient
CAS Number: 3002440-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR